CAS RN 10519-20-7
IUPAC Name:
ethyl oct-2-ynoate
Synonyms:
ethyl oct-2-ynoate
Molecular Weight (g/mol):
168.24
Molecular Formula:
C10H16O2
InChi Key:
QPMDWYXUSMRVKT-UHFFFAOYSA-N
SMILES:
CCCCCC#CC(=O)OCC
1
–
1
of
1
results
1
Ethyl 2-Octynoate, TCI America™
CAS: 10519-20-7 Molecular Formula: C10H16O2 Molecular Weight (g/mol): 168.24 MDL Number: MFCD00015286 InChI Key: QPMDWYXUSMRVKT-UHFFFAOYSA-N Synonym: ethyl 2-octynoate,2-octynoic acid, ethyl ester,2-octynoic acid ethyl ester,ethyl 2-octynate,ethyl heptynecarbonate,unii-xth780u0n3,ethyl heptine carbonate,ethyl n-oct-2-ynoate,ethyl heptyne carboxylate,2-octynoic acid, ethylester PubChem CID: 66338 IUPAC Name: ethyl oct-2-ynoate SMILES: CCCCCC#CC(=O)OCC