Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-Octynoate, TCI America™
SDP

Catalog No. O025325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O025325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O025325ML Supplier TCI America Supplier No. O025325ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10519-20-7
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00015286
InChI Key QPMDWYXUSMRVKT-UHFFFAOYSA-N
Synonym ethyl 2-octynoate, 2-octynoic acid, ethyl ester, 2-octynoic acid ethyl ester, ethyl 2-octynate, ethyl heptynecarbonate, unii-xth780u0n3, ethyl heptine carbonate, ethyl n-oct-2-ynoate, ethyl heptyne carboxylate, 2-octynoic acid, ethylester
PubChem CID 66338
IUPAC Name ethyl oct-2-ynoate
SMILES CCCCCC#CC(=O)OCC

Specifications

Color Colorless
Boiling Point 85°C
Quantity 25 mL
Formula Weight 168.24
Physical Form Liquid
Chemical Name or Material Ethyl 2-Octynoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.