accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1052707-07-9

IUPAC Name: 1,2-bis(4-nitrophenyl)ethane-1,2-diamine dihydrochloride
Synonyms: 1,2-bis(4-nitrophenyl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 375.21
Molecular Formula: C14H16Cl2N4O4
InChi Key: PDPYGNJVCKPVGM-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(C(N)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)