accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105365-50-2

IUPAC Name: (4-hexylphenyl)boronic acid
Synonyms: 4-hexylphenylboronic acid
Molecular Weight (g/mol): 206.09
Molecular Formula: C12H19BO2
InChi Key: CFYBKGAFWFRFRJ-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)