Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hexylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. H14895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1489-5G 5 g
H14891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1489-5G Supplier TCI America Supplier No. H14895G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 105365-50-2
Molecular Formula C12H19BO2
MDL Number MFCD04039029
InChI Key CFYBKGAFWFRFRJ-UHFFFAOYSA-N
PubChem CID 4197864
IUPAC Name (4-hexylphenyl)boronic acid

Specifications

Color White
Quantity 5 g
Synonym 4-hexylphenyl boronic acid, 4-n-hexylphenylboronic acid, 4-hex-1-yl benzeneboronic acid, 1-borono-4-hex-1-yl benzene, 4-n-hexylbenzeneboronic acid
SMILES CCCCCCC1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 206.09
Formula Weight 206.09
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.