CAS RN 1053655-59-6
IUPAC Name:
1-(oxan-2-yl)-1H-indazol-6-amine
Synonyms:
1-(oxan-2-yl)indazol-6-amine
Molecular Weight (g/mol):
217.27
Molecular Formula:
C12H15N3O
InChi Key:
BVMGAUOJVUEFSV-UHFFFAOYNA-N
SMILES:
NC1=CC=C2C=NN(C3CCCCO3)C2=C1
1
–
1
of
1
results
1
Sigma Aldrich 6-Amino-1-(tetrahydropyranyl)-1H-indazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.