accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHNOOHBr

CAS RN 1053659-26-9

IUPAC Name: 2-(2-bromophenyl)-1H-1,3-benzodiazole-6-carboxylic acid
Synonyms: 2-(2-bromophenyl)-3H-1,3-benzodiazole-5-carboxylic acid
Molecular Weight (g/mol): 317.14
Molecular Formula: C14H9BrN2O2
InChi Key: VDQXOCQBIBFAIF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results