accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10543-60-9

IUPAC Name: 1,3,4,6-tetraacetyl-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
Synonyms: 1,3,4,6-tetraacetyl-dihydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
Molecular Weight (g/mol): 310.27
Molecular Formula: C12H14N4O6
InChi Key: KIHGYZTVBURVBA-UHFFFAOYSA-N
SMILES: CC(=O)N1C2C(N(C(C)=O)C1=O)N(C(C)=O)C(=O)N2C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)