Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N',N'',N'''-Tetraacetylglycoluril, +98%, Thermo Scientific™

Catalog No. AC183841000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC183841000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC183841000 Supplier Thermo Scientific Chemicals Supplier No. 183841000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 10543-60-9
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Molecular Formula C12H14N4O6
MDL Number MFCD00022618
Quantity 100g
Beilstein 26, 443
Fieser 01,563
Synonym n,n',n,n'-tetraacetylglycoluril, 1,3,4,6-tetraacetyltetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, imidazo 4,5-d imidazole-2,5 1h,3h-dione, 1,3,4,6-tetraacetyltetrahydro, n,n',n,n'-tetraacetylacetyleneurea, n,n',n,n'-tetraacetylglycouril, 1,3,4,6-tetraacetyl-dihydro-1,3 diazolo 4,5-d imidazole-2,5-dione, tetraacetylglycoluril, tetraacetylglykoluril
InChI Key KIHGYZTVBURVBA-UHFFFAOYSA-N
SMILES CC(=O)N1C2C(N(C(C)=O)C1=O)N(C(C)=O)C(=O)N2C(C)=O
IUPAC Name 1,3,4,6-tetraacetyl-octahydro-[1,3]diazolo[4,5-d]imidazole-2,5-dione
Molecular Weight (g/mol) 310.27
PubChem CID 82717
Formula Weight 310.26
Chemical Name or Material N,N',N'',N'''-Tetraacetylglycoluril, 98+%
Show More Show Less
EINECSNumber 234-124-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.