accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CH3CCH3OHHO

CAS RN 10547-96-3

IUPAC Name: 2,2-bis(2-methylpropyl)propane-1,3-diol
Synonyms: 2,2-bis(2-methylpropyl)propane-1,3-diol
Molecular Weight (g/mol): 188.31
Molecular Formula: C11H24O2
InChi Key: PTRCHMOHGGDNIJ-UHFFFAOYSA-N
SMILES: CC(C)CC(CO)(CO)CC(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results