Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diisobutyl-1,3-propanediol 98.0+%, TCI America™
SDP

Catalog No. D27775G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27775G 5 g
D277725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27775G Supplier TCI America Supplier No. D27775G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10547-96-3
Molecular Formula C11H24O2
Molecular Weight (g/mol) 188.311
MDL Number MFCD00986394
InChI Key PTRCHMOHGGDNIJ-UHFFFAOYSA-N
Synonym 2,2-Dihydroxy-2,2-diisobutylpropane
PubChem CID 3985625
IUPAC Name 2,2-bis(2-methylpropyl)propane-1,3-diol
SMILES CC(C)CC(CC(C)C)(CO)CO

Specifications

Melting Point 77°C
Color White
Boiling Point 160°C
Quantity 5 g
Formula Weight 188.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2-Diisobutyl-1,3-propanediol
EINECSNumber (2)-0240
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.