accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOO

CAS RN 10551-58-3

IUPAC Name: (5-formylfuran-2-yl)methyl acetate
Synonyms: (5-formylfuran-2-yl)methyl acetate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: QAVITTVTXPZTSE-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=CC=C(O1)C=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)