Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Acetoxymethyl-2-furaldehyde, Technical Grade, Thermo Scientific™

Catalog No. AAA1182006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1182006 5 g
AAA1182003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1182006 Supplier Thermo Scientific Chemicals Supplier No. A1182006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10551-58-3
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00003233
InChI Key QAVITTVTXPZTSE-UHFFFAOYSA-N
Synonym 5-acetoxymethyl-2-furaldehyde, 5-formylfuran-2-yl methyl acetate, 5-acetoxymethylfurfural, 5-acetoxymethyl furfural, 5-formylfurfuryl acetate, 5-formyl-2-furyl methyl acetate, unii-5hh6180xyd, 5-acetoxymethyl furfural amf, 2-furancarboxaldehyde, 5-acetyloxy methyl
PubChem CID 66349
IUPAC Name (5-formylfuran-2-yl)methyl acetate
SMILES CC(=O)OCC1=CC=C(O1)C=O

Specifications

Melting Point 53°C to 55°C
Boiling Point 122°C (2 mmHg)
Flash Point 108°C (226°F)
Quantity 5 g
Beilstein 139154
Formula Weight 168.15
Grade Technical
Chemical Name or Material 5-Acetoxymethyl-2-furaldehyde
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.