accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFFFFFFFFH2NNH2HClHCl

CAS RN 1055301-07-9

IUPAC Name: 1,2-bis(2,3,4,5,6-pentafluorophenyl)ethane-1,2-diamine dihydrochloride
Synonyms: 1,2-bis(2,3,4,5,6-pentafluorophenyl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 465.12
Molecular Formula: C14H8Cl2F10N2
InChi Key: JXHUHXWIKZGKIE-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(C(N)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)