Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-(+)-1,2-Bis(2,3,4,5,6-pentafluorophenyl)-1,2-ethanediamine dihydrochloride (cont. 10-20wt% mono HCl), 95%, ee 99%, Thermo Scientific™

Catalog No. AAH25977MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25977MD 250 mg
AAH2597703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25977MD Supplier Thermo Scientific Chemicals Supplier No. H25977MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1055301-07-9
Molecular Formula C14H8Cl2F10N2
Molecular Weight (g/mol) 465.12
MDL Number MFCD09265309,MFCD09953690
InChI Key JXHUHXWIKZGKIE-UHFFFAOYNA-N
Synonym 1r,2r-1,2-bis 2,3,4,5,6-pentafluorophenyl ethane-1,2-diamine dihydrochloride, r,r-1,2-bis 2,3,4,5-pentafluorophenyl-1,2-ethanediamine dihydrochloride, ee, r,r-+-1,2-bis 2,3,4,5,6-pentafluorophenyl-1,2-ethanediamine dihydrochloride cont. 10-20wt% mono hcl
PubChem CID 73995081
IUPAC Name (1R,2R)-1,2-bis(2,3,4,5,6-pentafluorophenyl)ethane-1,2-diamine;dihydrochloride
SMILES Cl.Cl.NC(C(N)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Melting Point >200°C
Quantity 250 mg
Optical Rotation +28° (c=1 in Methanol)
Formula Weight 465.13
Percent Purity 95%
Assay ee 99%, (cont. 10 to 20wt% mono HCl)
Chemical Name or Material (R,R)-(+)-1,2-Bis(2,3,4,5,6-pentafluorophenyl)-1,2-ethanediamine dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.