CAS RN 1055301-12-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R,R)-1,2-Bis(4-cyanophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™
CAS: 1055301-12-6 Molecular Formula: C16H14N4·2HCl MDL Number: MFCD09265296