accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1055301-17-1

IUPAC Name: 1,2-bis(naphthalen-1-yl)ethane-1,2-diamine dihydrochloride
Synonyms: 1,2-bis(naphthalen-1-yl)ethane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 385.33
Molecular Formula: C22H22Cl2N2
InChi Key: SHNGCXWOHADIKG-UHFFFAOYNA-N
SMILES: Cl.Cl.NC(C(N)C1=C2C=CC=CC2=CC=C1)C1=CC=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)