CAS RN 1055888-89-5
IUPAC Name:
3-(di-tert-butylphosphanyl)propane-1-sulfonic acid
Synonyms:
3-(di-tert-butylphosphanyl)propane-1-sulfonic acid
Molecular Weight (g/mol):
268.35
Molecular Formula:
C11H25O3PS
InChi Key:
JPNPRWMRUCIEMN-UHFFFAOYSA-N
SMILES:
CC(C)(C)P(CCCS(O)(=O)=O)C(C)(C)C
1
–
1
of
1
results
1
Promotions Available
3-(Di-tert-butylphosphonium)propane sulfonate, 97%
3-(Di-tert-butylphosphonium)propane sulfonate, 97%
CAS: 1055888-89-5 Molecular Formula: C11H25O3PS Molecular Weight (g/mol): 268.352 MDL Number: MFCD12546062 InChI Key: JPNPRWMRUCIEMN-UHFFFAOYSA-N Synonym: 3-di-t-butylphosphonium propane sulfonate,3-di-tert-butylphosphino propane-1-sulfonic acid,dtbpps,di-t-butyl 3-sulfonatopropyl phosphine,3-di-tert-butylphosphonium propane sulfonate,3-di-tert-butylphosphanyl propane-1-sulfonic acid PubChem CID: 24949188 IUPAC Name: 3-ditert-butylphosphanylpropane-1-sulfonic acid SMILES: CC(C)(C)P(CCCS(=O)(=O)O)C(C)(C)C