accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOHNH2(E/Z)

CAS RN 1055926-31-2

IUPAC Name: N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide
Synonyms: N'-hydroxy-2-(4-methoxyphenyl)ethanimidamide
Molecular Weight (g/mol): 180.21
Molecular Formula: C9H12N2O2
InChi Key: QIACLRIKXICINP-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(N)=NO)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results