accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3IH3CHO

CAS RN 10570-67-9

IUPAC Name: 4-iodo-2,6-dimethylphenol
Synonyms: 4-iodo-2,6-dimethylphenol
Molecular Weight (g/mol): 248.06
Molecular Formula: C8H9IO
InChi Key: HUUNIMCCAGNBDF-UHFFFAOYSA-N
SMILES: CC1=CC(I)=CC(C)=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results