Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-2,6-dimethylphenol 97.0+%, TCI America™
SDP

Catalog No. I06005G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06005G Supplier TCI America Supplier No. I06005G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10570-67-9
Molecular Formula C8H9IO
Molecular Weight (g/mol) 248.06
MDL Number MFCD00094415
InChI Key HUUNIMCCAGNBDF-UHFFFAOYSA-N
Synonym 2,6-dimethyl-4-iodophenol, 4-iodo-2,6-dimethyl-phenol, 4-iodo-2,6-xylenol, acmc-2098hg, phenol,4-iodo-2,6-dimethyl, huunimccagnbdf-uhfffaoysa, phenol, 4-iodo-2,6-dimethyl
PubChem CID 555318
IUPAC Name 4-iodo-2,6-dimethylphenol
SMILES CC1=CC(I)=CC(C)=C1O

Specifications

Melting Point 102°C
Quantity 5 g
Formula Weight 248.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-2,6-dimethylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.