accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHSOOO

CAS RN 10580-19-5

IUPAC Name: 4-hydroxybenzene-1-sulfonate
Synonyms: 4-hydroxybenzenesulfonate
Molecular Weight (g/mol): 173.16
Molecular Formula: C6H5O4S
InChi Key: FEPBITJSIHRMRT-UHFFFAOYSA-M
SMILES: OC1=CC=C(C=C1)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)