accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPNPNPNH3CH3CCH3CH3CH3

CAS RN 105809-15-2

IUPAC Name: hexamethyl-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Synonyms: hexamethyl-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Molecular Weight (g/mol): 225.15
Molecular Formula: C6H18N3P3
InChi Key: DEBZEVJNWCNATM-UHFFFAOYSA-N
SMILES: CP1(C)=NP(C)(C)=NP(C)(C)=N1
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results