Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ultramark∣r 1621, Mass Spec Std

Catalog No. AAL1669803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1669803 1 g
AAL16698MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1669803 Supplier Thermo Scientific Chemicals Supplier No. L1669803
  • $184.50
    $150.65 / Each of 1

    Save $33.85

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 105809-15-2 | C6H18N3P3 | 231.189 g/mol

Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications

Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

Solubility

Miscible with water.

Notes

Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105809-15-2
Molecular Formula C6H18N3P3
Molecular Weight (g/mol) 231.189
MDL Number MFCD00242454
InChI Key DEBZEVJNWCNATM-MZWXYZOWSA-N
PubChem CID 44119860
IUPAC Name 2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES CP1(=NP(=NP(=N1)(C)C)(C)C)C

Specifications

Density 1.8
Solubility Miscible with water
Type Mass Spectrometry Standards
Quantity 1 g
Formula Weight 2120
Grade Reference Material
Chemical Name or Material Ultramark™ 1621, Mass Spec Standard
Hazard Category H315-H319-H335
Hazard Statement GHS H StatementH315-H319-H335 Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.