accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONNOOH3CCH3CH3CH3BFFFF

CAS RN 105832-38-0

IUPAC Name: [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
Synonyms: [(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium tetrafluoroborate
Molecular Weight (g/mol): 301.05
Molecular Formula: C9H16BF4N3O3
InChi Key: YEBLHMRPZHNTEK-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (3)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (2)
  • (4)
  • (7)
  • (2)
  • (3)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)