Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSTU, Advanced ChemTech

Catalog No. 01339230
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-339-230 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-339-230 Supplier Advanced Chem Tech Supplier No. RT858525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Weight (g/mol) 301.05

Specifications

CAS 105832-38-0
Color White/Off-White
Quantity 25 g
Grade Biochemical
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.