accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOH

CAS RN 10595-09-2

IUPAC Name: 3-[(3-hydroxypropyl)sulfanyl]propan-1-ol
Synonyms: 3-[(3-hydroxypropyl)sulfanyl]propan-1-ol
Molecular Weight (g/mol): 150.24
Molecular Formula: C6H14O2S
InChi Key: QRQVZZMTKYXEKC-UHFFFAOYSA-N
SMILES: OCCCSCCCO
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results