accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3OH(E)

CAS RN 106-24-1

IUPAC Name: (2E)-3,7-dimethylocta-2,6-dien-1-ol
Synonyms: geraniol t-geraniol β-geraniol geraniol (natural) citrol lemonol epa pesticide chemical code 597501 geraniol extra
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: GLZPCOQZEFWAFX-JXMROGBWSA-N
SMILES: CC(C)=CCC\C(C)=C\CO
Molecular Weight (g/mol) 
  • (10)
  • (6)
Percent Purity 
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
Boiling Point 
  • (3)
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (1)
  • (2)
  • (3)
  • (8)
  • (10)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)