Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Geraniol

Catalog No. 16333325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-333-325G 25 g
16-333-3100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-333-325G Supplier Sigma Aldrich Supplier No. 16333325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Geraniol undergoes oxidation with molecular oxygen in supercritical carbon dioxide catalyzed by chromium containing mesoporous molecular sieve to yield citral.geraniol was used in field evaluation of synthetic herbivore-induced plant volatiles as attractants to beneficial insects
  • It was used to evaluate the tumor-suppressive potency of isoprenoids in vitro and in vivo.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • 163333.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 106-24-1
Density 0.879 g/mL (at 20°C (literature))
Molecular Formula C10H18O
Linear Formula (CH3)2 C=CHCH2CH2C(CH3)=CHCH2OH
MDL Number MFCD00002917
Boiling Point 229°C to 230°C (lit.)
Synonym trans-3,7-Dimethyl-2,6-octadien-1-ol
Molecular Weight (g/mol) 154.25
Quantity 25 g
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.