CAS RN 106-51-4
p-Benzoquinone, 98+%
CAS: 106-51-4 Molecular Formula: C6H4O2 Molecular Weight (g/mol): 108.096 MDL Number: MFCD00001591 InChI Key: AZQWKYJCGOJGHM-UHFFFAOYSA-N Synonym: p-benzoquinone,benzoquinone,quinone,1,4-benzoquinone,p-quinone,chinone,2,5-cyclohexadiene-1,4-dione,cyclohexadienedione,para-quinone,1,4-benzoquine PubChem CID: 4650 ChEBI: CHEBI:16509 IUPAC Name: cyclohexa-2,5-diene-1,4-dione SMILES: C1=CC(=O)C=CC1=O
1,4-Benzoquinone, 98+%
CAS: 106-51-4 Molecular Formula: C6H4O2 Molecular Weight (g/mol): 108.1 MDL Number: MFCD00001591 InChI Key: AZQWKYJCGOJGHM-UHFFFAOYSA-N Synonym: p-benzoquinone,benzoquinone,quinone,1,4-benzoquinone,p-quinone,chinone,2,5-cyclohexadiene-1,4-dione,cyclohexadienedione,para-quinone,1,4-benzoquine PubChem CID: 4650 ChEBI: CHEBI:16509 IUPAC Name: cyclohexa-2,5-diene-1,4-dione SMILES: C1=CC(=O)C=CC1=O
p-Benzoquinone, ≥99.5% (HPLC), MilliporeSigma™ Supelco™
MDL Number: MFCD00001591 Synonym: Quinone
1,4-Benzoquinone 97.0+%, TCI America™
CAS: 106-51-4 Molecular Formula: C6H4O2 Molecular Weight (g/mol): 108.096 MDL Number: MFCD00001591 InChI Key: AZQWKYJCGOJGHM-UHFFFAOYSA-N Synonym: p-benzoquinone,benzoquinone,quinone,1,4-benzoquinone,p-quinone,chinone,2,5-cyclohexadiene-1,4-dione,cyclohexadienedione,para-quinone,1,4-benzoquine PubChem CID: 4650 ChEBI: CHEBI:16509 IUPAC Name: cyclohexa-2,5-diene-1,4-dione SMILES: C1=CC(=O)C=CC1=O
1,4-Benzoquinone 98.0+%, TCI America™
CAS: 106-51-4 Molecular Formula: C6H4O2 Molecular Weight (g/mol): 108.096 MDL Number: MFCD00001591 InChI Key: AZQWKYJCGOJGHM-UHFFFAOYSA-N Synonym: p-benzoquinone,benzoquinone,quinone,1,4-benzoquinone,p-quinone,chinone,2,5-cyclohexadiene-1,4-dione,cyclohexadienedione,para-quinone,1,4-benzoquine PubChem CID: 4650 ChEBI: CHEBI:16509 IUPAC Name: cyclohexa-2,5-diene-1,4-dione SMILES: C1=CC(=O)C=CC1=O
Sigma Aldrich (-)-Pi Reagent
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich P-Benzoquinone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
1,4-Benzoquinone, SPEX CertiPrep™
CAS: 106-51-4 Molecular Formula: C6H4O2 Molecular Weight (g/mol): 108.096 InChI Key: AZQWKYJCGOJGHM-UHFFFAOYSA-N PubChem CID: 4650 ChEBI: CHEBI:16509 IUPAC Name: cyclohexa-2,5-diene-1,4-dione SMILES: C1=CC(=O)C=CC1=O