Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Benzoquinone, ≥99.5% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111019219 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9219 25 g
11-101-8250 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9219 Supplier MilliporeSigma Supelco Supplier No. 1230925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00001591
Synonym Quinone

Specifications

CAS 106-51-4
Melting Point 112°C to 114°C; 113°C to 115°C
Ignition Point 815°F
Linear Formula C6H4(=O)2
Quantity 25 g
Molecular Weight (g/mol) 108.09
Vapor Density 3.73 (vs. air)
Vapor Pressure 0.1 mmHg (25°C)
Formula Weight 108.09
Percent Purity ≥99.5% (HPLC)
Grade Not Available
Physical Form Powder
Show More Show Less
RTECSNumber DK2625000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.