accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 106-79-6

IUPAC Name: 1,10-dimethyl decanedioate
Synonyms: decanedioic acid, dimethyl ester
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: ALOUNLDAKADEEB-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCC(=O)OC
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)