Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethyl sebacate

Catalog No. 223115500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-311-5500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-311-5500G Supplier Sigma Aldrich Supplier No. 223115500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethyl sebacate

Specifications

CAS 106-79-6
Melting Point 29°C to 31°C
Density 0.988 g/mL (at 25°C)
Boiling Point 158°C
Molecular Formula C12H22O4
Linear Formula CH3OCO(CH2)8 COOCH3
Quantity 500 g
MDL Number MFCD00008472
Synonym Decanedioic acid 1,10-dimethyl ester; Dimethyl decanedioate; NSC 9415; Sebacic acid dimethyl ester
Molecular Weight (g/mol) 230.3
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.