accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106-86-5

CH2O

CAS RN 106-86-5

IUPAC Name: 3-ethenyl-7-oxabicyclo[4.1.0]heptane
Synonyms: 4-vinylcyclohexene dioxide
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: SLJFKNONPLNAPF-UHFFFAOYNA-N
SMILES: C=CC1CCC2OC2C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)