accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOH3COCH3

CAS RN 10601-80-6

IUPAC Name: ethyl 3,3-diethoxypropanoate
Synonyms: ethyl 3,3-diethoxypropanoate
Molecular Weight (g/mol): 190.24
Molecular Formula: C9H18O4
InChi Key: SIALOQYKFQEKOG-UHFFFAOYSA-N
SMILES: CCOC(CC(=O)OCC)OCC
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (3)
  • (4)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)