accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 10606-42-5

IUPAC Name: ethyl 3-methoxypropanoate
Synonyms: ethyl 3-methoxypropanoate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: IJUHLFUALMUWOM-UHFFFAOYSA-N
SMILES: CCOC(=O)CCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results