CAS RN 1062169-56-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ mTOR Kinase Inhibitor II, WYE-354
The mTOR Kinase Inhibitor II, WYE-354, also referenced under CAS 1062169-56-5, controls the biological activity of mTOR. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.