accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 106224-36-6

IUPAC Name: palladium(2+) bis(2,2-dimethylpropanoate)
Synonyms: palladium(2+) dipivalate
Molecular Weight (g/mol): 308.67
Molecular Formula: C10H18O4Pd
InChi Key: PAGZTSLSNQZYEV-UHFFFAOYSA-L
SMILES: [Pd++].CC(C)(C)C([O-])=O.CC(C)(C)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results