Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Azidotoluene Solution

Catalog No. 72746610ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-746-610ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 72-746-610ML Supplier Sigma Aldrich Supplier No. 72746610ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Azidotoluene is an aromatic azide generally used in copper(I)-catalyzed azide-alkyne cycloaddition reactions.4-Azidotoluene solution may be used for the functionalization at the axial position of boron subphthalocyanines with a terminal acetylene functionality via copper(I)-catalyzed azide-alkyne cycloaddition in the presence of Hunig'sas base.

Specifications

CAS 106224-36-6
Melting Point 230°C to 232°C
Molecular Formula C10H18O4Pd
Linear Formula C10H18O4Pd
MDL Number MFCD16038138
Quantity 500 mg
Synonym 2,2-Dimethylpropanoic acid palladium(II) salt
Molecular Weight (g/mol) 308.67 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.