accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H3CCl

CAS RN 106315-37-1

IUPAC Name: ethyl 3-chloro-2,2-dimethylpropanoate
Synonyms: ethyl 3-chloro-2,2-dimethylpropanoate
Molecular Weight (g/mol): 164.63
Molecular Formula: C7H13ClO2
InChi Key: NEVHNBRNUWSFSB-UHFFFAOYSA-N
SMILES: CCOC(=O)C(C)(C)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results