accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPClCl(E/Z)

CAS RN 106423-29-4

IUPAC Name: triphenyl[4-(triphenylphosphaniumyl)but-2-en-1-yl]phosphanium dichloride
Synonyms: triphenyl[4-(triphenylphosphaniumyl)but-2-en-1-yl]phosphanium dichloride
Molecular Weight (g/mol): 649.58
Molecular Formula: C40H36Cl2P2
InChi Key: OOACLPDZHDFCQP-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].C(C=CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results