Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-2-Butene-1,4-bis(triphenylphosphonium Chloride) 95.0+%, TCI America™
SDP

Catalog No. B22865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22865G Supplier TCI America Supplier No. B22865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106423-29-4
Molecular Formula C40H36Cl2P2
Molecular Weight (g/mol) 649.576
MDL Number MFCD00031713
InChI Key OOACLPDZHDFCQP-UHFFFAOYSA-L
Synonym 2-butene-1,4-bis triphenylphosphonium chloride, trans-2-butene-1,4-bis triphenylphosphoniumchloride, but-2-ene-1,4-diyl bis triphenylphosphanium dichloride
PubChem CID 70700154
IUPAC Name triphenyl(4-triphenylphosphaniumylbut-2-enyl)phosphanium;dichloride
SMILES C1=CC=C(C=C1)[P+](CC=CC[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6.[Cl-].[Cl-]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 649.58
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2-Butene-1,4-bis(triphenylphosphonium Chloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.