accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NPOHOHO

CAS RN 1066-51-9

IUPAC Name: (aminomethyl)phosphonic acid
Synonyms: aminomethylphosphonic acid (1-aminomethyl)phosphonic acid epa pesticide chemical code 207800 caswell no. 037C AMeP
Molecular Weight (g/mol): 111.04
Molecular Formula: CH6NO3P
InChi Key: MGRVRXRGTBOSHW-UHFFFAOYSA-N
SMILES: NCP(O)(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (4)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (6)
  • (5)
  • (5)
  • (2)
  • (8)
  • (4)
  • (3)
  • (3)
  • (2)
  • (3)