Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Aminomethyl)phosphonic acid

Catalog No. 324817250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-481-7250MG 250 mg
32-481-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-481-7250MG Supplier Sigma Aldrich Supplier No. 324817250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(Aminomethyl)phosphonic acid is a metabolite of amitrole and glyphosate herbicides and was analyzed by HPLC using an electrochemical detector.(Aminomethyl)phosphonic acid was used to develop a novel kind of immobilized metal affinity chromatography column based on organic-inorganic hybrid silica monolith.

Specifications

CAS 1066-51-9
Molecular Formula CH6NO3P
Linear Formula NH2CH2P(O)(OH)2
MDL Number MFCD00008105
Quantity 250 mg
Synonym AMPA
Molecular Weight (g/mol) 111.04
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.