accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOCH3OCH3OONOO

CAS RN 1069137-73-0

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2,5,7,8-tetramethyl-6-(oxan-2-yloxy)-3,4-dihydro-2H-1-benzopyran-2-carboxylate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2,5,7,8-tetramethyl-6-(oxan-2-yloxy)-3,4-dihydro-1-benzopyran-2-carboxylate
Molecular Weight (g/mol): 431.49
Molecular Formula: C23H29NO7
InChi Key: WDEFWOPTRFWZLM-UHFFFAOYNA-N
SMILES: CC1=C(OC2CCCCO2)C(C)=C2CCC(C)(OC2=C1C)C(=O)ON1C(=O)CCC1=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results