Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinimidyl (2R)-6-(Tetrahydro-2H-pyran-2-yloxy)-2,5,7,8-tetramethylchroman-2-carboxylate, TCI America™
SDP

Catalog No. S059950MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S059950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S059950MG Supplier TCI America Supplier No. S059950MG

Chemical Identifiers

CAS 1069137-73-0
Molecular Formula C23H29NO7
Molecular Weight (g/mol) 431.49
MDL Number MFCD11042683
InChI Key WDEFWOPTRFWZLM-UHFFFAOYNA-N
Synonym (2R)-6-(Tetrahydro-2H-pyran-2-yloxy)-2,5,7,8-tetramethylchroman-2-carboxylic Acid Succinimidyl Ester, NPCA
PubChem CID 44630342
IUPAC Name 2,5-dioxopyrrolidin-1-yl 2,5,7,8-tetramethyl-6-(oxan-2-yloxy)-3,4-dihydro-2H-1-benzopyran-2-carboxylate
SMILES CC1=C(OC2CCCCO2)C(C)=C2CCC(C)(OC2=C1C)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 174°C
Color White
Quantity 50 mg
Formula Weight 431.49
Physical Form Crystalline Powder
Chemical Name or Material Succinimidyl (2R)-6-(Tetrahydro-2H-pyran-2-yloxy)-2,5,7,8-tetramethylchroman-2-carboxylate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.