accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClN

CAS RN 107-14-2

IUPAC Name: 2-chloroacetonitrile
Synonyms: chloroacetonitrile
Molecular Weight (g/mol): 75.5
Molecular Formula: C2H2ClN
InChi Key: RENMDAKOXSCIGH-UHFFFAOYSA-N
SMILES: ClCC#N
Molecular Weight (g/mol) 
  • (4)
  • (3)
Boiling Point 
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)