Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloroacetonitrile

Catalog No. C196515G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C196515G 5 g
C19651500G 500 g
C19651100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C196515G Supplier Sigma Aldrich Supplier No. C196515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Chloroacetonitrile can be used:
  • For the synthesis of ionic liquids containing nitrile functionalized imidazolium salts
  • For the synthesis of cyanomethyl dodecyl trithiocarbonate, a reversible addition-fragmentation chain transfer (raft) agent
  • As an alkylating reagent for the synthesis of derivatives of triazolyl thiazoles to be used as anti-tuberculosis agents
  • As a reagent in photo meerwein addition reaction for amino-arylation of alkenes
  • As an esterifying agent in the synthesis of a cytotoxic natural product: apicularen a

Specifications

CAS 107-14-2
Density 1.193 g/mL (at 25°C (literature))
Boiling Point 124°C to 126°C (lit.)
Molecular Formula C2H2ClN
Refractive Index n20/D 1.422 (literature)
Linear Formula ClCH2CN
MDL Number MFCD00001885
Quantity 5 g
Molecular Weight (g/mol) 75.5
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.