accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 107-22-2

IUPAC Name: oxaldehyde
Synonyms: glyoxal oxal glyoxal aldehyde biformal biformyl oxalaldehyde Oxal ccris 952 aerotex glyoxal 40 diformyl
Molecular Weight (g/mol): 58.04
Molecular Formula: C2H2O2
InChi Key: LEQAOMBKQFMDFZ-UHFFFAOYSA-N
SMILES: O=CC=O
Molecular Weight (g/mol) 
  • (8)
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (4)
Physical Form 
  • (2)
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (4)
  • (2)
  • (8)
  • (8)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (2)