Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Glyoxal Solution

Catalog No. 1284655G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
3 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-846-55G 5 g
12-846-5100G 100 g
12-846-51KG 1 kg
12-846-53KG 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Warning
Catalog No. 12-846-55G Supplier Sigma Aldrich Supplier No. 1284655G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is 40wt.% solution of glyoxal in water. Glyoxal is a linear aliphatic dialdehyde containing two aldehyde groups. It participates in the synthesis of glyoxylic acid. It is highly reactive in nature. It can be prepared by oxidizing ethanol or acetaldehyde with nitric acid. It is widely employed in textile and paper industry.Glyoxal solution has been used as a crosslinking agent during the synthesis of DNA-containing nanoparticles in a study. It has been used as derivatizing reagent, in the precolumn fluorescence derivatization of N-terminal tryptophan containing peptides which can be separated via high performance liquid chromatography. It may also be used in the synthesis of glyoxal bis(4-phenyl-3-thiosemicarbazones)solution, which can be used in the spectrophotometric determination of palladium in catalysts.This form of glyoxal is composed of 3 moles of glyoxal and 2 moles of water in a relatively stable configuration.

Specifications

CAS 107-22-2
Density 1.265 g/mL at 25°C
Molecular Formula C2H2O2
Linear Formula OHCCHO
Refractive Index n20/D 1.4087
Quantity 5 g
MDL Number MFCD00006957
Synonym Ethanedial; Oxalaldehyde
Molecular Weight (g/mol) 58.04 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.